missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 3 Knuckle Coated Cut-Resistant Gloves

Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 3 Knuckle Coated Cut-Resistant Gloves are for industrial tasks that require protection from abrasions, cuts and punctures

$145.88 - $145.90

Specifications

Coating Material Textured
Color Black, Orange
Cut Resistance Rating 3
Material Dyneema™, Nylon, Lycra™
For Use With (Application) Metal fabrication, Glass handling, automotive assembly, stamping and assembly
View More Specs

Products 5
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity  
19-005-415 View Documents Kimberly-Clark™ Professional
KC38647
8.4 in. 21.3cm Small Case for $145.90
 
19-005-416 View Documents Kimberly-Clark™ Professional
KC38648
8.9 in. 22.6cm Medium Case for $145.90
 
19-005-419 View Documents Kimberly-Clark™ Professional
KC38651
10 in. 25.4cm 2X-Large Case for $145.88
 
19-005-418 View Documents Kimberly-Clark™ Professional
KC38650
9.6 in. 24.3cm X-Large Case for $145.88
 
19-005-417 View Documents Kimberly-Clark™ Professional
KC38649
9.3 in. 23.6cm Large Case for $145.88
 
Description

Description

  • Offer EN388 Level 3 / ANSI Level 2 Cut Resistance and CPPT = 512 grams
  • EN388 Level 4 abrasion and Level 4 tear resistance for superior durability
  • Certified to meet the EN420 standards, including qualities such as glove design and construction, water resistance, innocuousness, and comfort
  • Bright orange fiber aids in overall safety compliance and awareness of how close a hand is in proximity to potential dangers
  • Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and stretch fiber, which provides both cut resistance and a comfortable fit
  • Polyurethane coating enhances dry and oil grip and hides dirt for longer wear
  • Dark coating helps hide dirt for longer wear
  • Each pair has a dedicated right and left glove for a snug fit
  • Knuckle coating helps keep hands and gloves protected from liquids and oils to enforce compliance and improve glove longevity
Specifications

Specifications

Textured
3
Metal fabrication, Glass handling, automotive assembly, stamping and assembly
Black, Orange
Dyneema™, Nylon, Lycra™
Knit