missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ G60 Knuckle Coated High-Visibility Cut-Resistant Gloves
High-visibility cut-resistant gloves with polyurethane coating are ideal for working in environments that pose a risk of minor cuts and abrasions.
Supplier: Kimberly-Clark Professional™ 38650
Description
- EN 388 Level 3 / ANSI Level 2 Cut Resistance
- EN 388 Level 4 abrasion and Level 4 tear resistance
- Certified to meet the EN 420 standards
- Bright orange fiber aids in overall safety compliance and awareness of how close a hand is in proximity to potential dangers
- Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and stretch fiber, which provides both cut resistance and a comfortable fit
- Polyurethane coating enhances dry and oil grip and hides dirt for longer wear
- Dark coating helps hide dirt for longer wear
- Each pair has a dedicated right and left glove for a snug fit
- Knuckle coating helps keep hands and gloves protected from liquids and oils to enforce compliance and improve glove longevity
Specifications
ANSI 4 | |
Black, Orange | |
Dyneema™, Nylon, Lycra | |
X-Large | |
Metal fabrication, Glass handling, automotive assembly, stamping and assembly | |
38650 | |
9.6 in. |
EN 420 | |
ANSI A3 | |
Cut-resistant Gloves | |
EN 388 | |
Knit | |
Kleenguard™ G60 | |
24.3 cm |