missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 2 Polyurethane Coated Cut Resistant Gloves

Great choice for working environments that pose a risk of minor cuts and abrasions

$88.63 - $88.63

Specifications

Coating Material Polyurethane
Color Gray, Yellow
Cut Resistance Rating Level 2
Material Kevlar™
For Use With (Application) Metal fabrication, Glass handling, automotive assembly, stamping and assembly
View More Specs

Alert:

This product contains natural rubber latex, which may cause allergic reactions. This product should not be worn by, or exposed to, individuals allergic to natural rubber latex. Safe use of this product on or by latex-sensitive individuals has no t been established.


Products 5
Catalog Number Mfr. No. Length (English) Length (Metric) Size Hem Color Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Size Hem Color Price Quantity  
19-169-400 View Documents Kimberly-Clark™ Professional
KC38635
8.661 in. 21.9cm Small Gray Case of 12 Each for $88.63
 
19-169-401 View Documents Kimberly-Clark™ Professional
KC38636
9.05 in. 22.9cm Medium Navy blue Case of 12 Pair for $88.63
 
19-169-402 View Documents Kimberly-Clark™ Professional
KC38637
9.44 in. 23.9cm Large Purple Case of 12 Pair for $88.63
 
19-169-403 View Documents Kimberly-Clark™ Professional
KC38638
9.84 in. 24.9cm X-Large Black Case of 12 Pair for $88.63
 
19-169-404 View Documents Kimberly-Clark™ Professional
KC38639
10.2 in. 25.9cm 2X-Large Brown Case of 12 Pair for $88.63
 
Description

Description

  • Kevlar™/spandex (15 gauge) seamless knit yellow shell
  • Stretch Kevlar liner provides cut resistance
  • Comfortable fit
  • Great dexterity and oil grip
  • Polyurethane coated palm provides a barrier against oil penetration
  • Color coded overlock for sizing
  • Suitable for intermittent heat contact up to 100°C (212°F)
  • Color: gray and yellow

Applications:

Metal fabrication, glass handling, automotive and aerospace assembly, stamping and assembly

Compliance

ANSI™/EN 388 Level 2 cut resistance

Specifications

Specifications

Polyurethane
Level 2
Metal fabrication, Glass handling, automotive assembly, stamping and assembly
Gray, Yellow
Kevlar™