missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Jackson Saftey™ G60 Level 2 Knuckle Coated Cut-Resistant Gloves

Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 2 Knuckle Coated Cut-Resistant Gloves are ideal both dexterity and cut resistance for risk of minor cuts and abrasions

$99.42 - $99.42

Specifications

Coating Material Textured
Color Gray,Yellow
Cut Resistance Rating 2
Material Dupont™ Kevlar™ aramid fibers
For Use With (Application) Metal fabrication, Glass handling, automotive assembly, stamping and assembly
View More Specs

Products 5
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity  
19-005-412 View Documents Kimberly-Clark Professional™
KC38644
9.3 in. 23.6cm Large Case for $99.42
 
19-005-410 View Documents Kimberly-Clark Professional™
KC38642
8.4 in. 21.3cm Small Case for $99.42
 
19-005-411 View Documents Kimberly-Clark Professional™
KC38643
8.9 in. 22.6cm Medium Case for $99.42
 
19-005-414 View Documents Kimberly-Clark Professional™
KC38646
10 in. 25.4cm 2X-Large Case for $99.42
 
19-005-413 View Documents Kimberly-Clark Professional™
KC38645
9.6 in. 24.3cm X-Large Case for $99.42
 
Description

Description

  • ANSI Level 2 / EN388 Level 2 cut resistance (CPPT = 672 grams)
  • Suitable for intermittent heat contact up to 212°F
  • Liner offers comfort and polyurethane coating provides a barrier against oil penetration
  • Excellent dexterity for improved productivity
  • Each pair has a dedicated right and left glove, for a snug fit
Specifications

Specifications

Textured
2
Metal fabrication, Glass handling, automotive assembly, stamping and assembly
Gray,Yellow
Dupont™ Kevlar™ aramid fibers
Knit