Please login to your online account to display your discounted pricing
Kimberly-Clark™ Professional Classic™ Surgical Masks
$69.00 - $115.71
Specifications
Construction 3-ply construction
Material Fabric
Filter Efficiency ≥96% (BFE), ≥97% (PFE)
Pore Size 0.1 (PFE), 3 (BFE)μ
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Color Includes Size Price Quantity    
18-999-2887
kimberly-clark™ professional
KC48201
Blue Tie closures Universal Case of 300 Each for $115.71
19-166-998
kimberly-clark™ professional
KC48296
Pink, blue Tie closures Regular Case of 300 Each for $69.00
Description & Specifications
Specifications
Construction 3-ply construction
Material Fabric
Filter Efficiency ≥96% (BFE), ≥97% (PFE)
Pore Size 0.1 (PFE), 3 (BFE)μ
Sterile No
Style Pleated
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.