missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ KIMAX™ Weighing Bottles — With standard taper Stoppers
Tall, cylindrical form with standard taper stoppers
Supplier: DWK Life Sciences 151453060
Description
With standard taper Stoppers
- Plug stopper fits into internally ground body; has closed bottom
- Designed from Fed. Spec. NNN-B-1493, Type I, Style I
Specifications
Glass | |
2.36 in. | |
1.01 oz., 30mL | |
1.01 oz. | |
30 mm | |
Glass | |
30g | |
Cylindrical |
60 mm | |
Plug | |
30 mL | |
1.18 in. | |
Low Form | |
29/12 | |
0.06 lb. |