missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Weighing Bottles — With standard taper Outside Ground Caps

Tall, cylindrical form with standard taper caps

Manufacturer:  DWK Life Sciences K151464050

 View more versions of this product

Catalog No. 03-422F


 

Description

Description

Tall, cylindrical form with standard taper caps

With Outside standard taper Ground Caps

  • Caps protect bottle in case of tipping; have open bottom
  • Prevent material from touching cap edges, provide increased headroom
  • Designed from Fed. Spec. NNN-B-1493, Type I, Style II

Specifications

Specifications

1.52 oz., 45mL
1.52 oz.
1.57 in.
Tall Form and Regular
45/12
0.12 lb.
Cylindrical
45mL
Glass
40mm
1.96 in.
55g
50mm
Cap
Documents