missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Tall-Form Beakers, with spout

With spout for easy pouring.

Manufacturer:  Kimble™ K14030300

 View more versions of this product

Catalog No. 02-539-9



 

Specifications Description & Specifications

Specifications

11.8cm
300mL
ASTM, E960, Type III, USP Type I, ASTM E438, Type I, Class A
6.4cm
For performing titrations
25 to 250mL
25mL
12 Pack
Tall Flared Top
10.14 oz.
4.64 in.
Borosilicate Glass
2.51 in.
No
Yes
Spout
Matte finish marking
Dual white scale
10.14 oz., 300mL
Beaker
Accessories