missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Tall-Form Beakers, with spout

With spout for easy pouring.

Manufacturer:  DWK Life Sciences K14030600

 View more versions of this product

Catalog No. 02-539-11



 

Description

Description

  • Matte-finish marking spot
  • Graduated to indicate approximate volumes
  • Berzelius style with flared top, reinforcing bead

Compliance

Designed from ASTM™ E 960 Type III (with spout) and Type IV (without spout) requirements

Specifications

Specifications

20.28 oz.
3.14 in.
No
Yes
5.9 in.
Spout
Matte finish marking
Dual white scale
Beaker
Borosilicate Glass
ASTM, E960, Type III, USP Type I, ASTM E438, Type I, Class A
8cm
For performing titrations
50 to 500mL
15cm
50mL
6 Pack
Tall Flared Top
600mL
Documents
Accessories