missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Tall-Form Beakers, without spout

Without spout.

Manufacturer:  Kimble™ K14020200

 View more versions of this product

Catalog No. 10-310-7


 

Specifications Description & Specifications

Specifications

10.4cm
200mL
ASTM, E960, Type IV, USP Type I, ASTM E438, Type I, Class A
5.7cm
For performing titrations
25 to 125mL
25mL
12 Case
Tall Flared Top
6.76 oz.
4.09 in.
Borosilicate Glass
2.24 in.
No
Yes
Without Spout
Matte finish marking
Dual white scale
6.76 oz., 200mL
Beaker
Accessories