missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Tall-Form Beakers, without spout

Without spout.

Manufacturer:  Kimble™ K140201000

 View more versions of this product

Catalog No. 10-310-6


 

Specifications Description & Specifications

Specifications

18.8cm
1000mL
ASTM, E960, Type IV, USP Type I, ASTM E438, Type I, Class A
9cm
For performing titrations
50 to 950mL
50mL
6 Case
Tall Flared Top
33.81 oz.
7.4 in.
Borosilicate Glass
3.54 in.
No
Yes
Without Spout
Matte finish marking
Dual white scale
33.81 oz., 1000mL
Beaker
Accessories