missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Serialized Class A Cylinders with Single Metric Scale

Calibrated “To Deliver”

Manufacturer:  Kimble™ K2002650

 View more versions of this product

Catalog No. 08-548C



 

Specifications Description & Specifications

Specifications

Graduated 3 to 50mL
Borosilicate Glass
A
Provides an easy way for a laboratory to meet QC/QA requirements
Certificate of graduation accuracy
±0.25mL
1mL
190mm
ASTM E1272
50mL
7.48 in.
Single metric scale
1mL
Accessories