Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets
Autoclavable
$182.78 - $1,127.58
Specifications
Certifications/Compliance Certificate of graduation accuracy, ASTM E969
Class A
Material Borosilicate Glass
Product Type Serialized Volumetric Pipet
Disposable No
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Capacity (Metric) Price Quantity    
13-660V
kimble™
K3701012
18.11 in. 46cm 12mL Pack of 6 Each for $385.98
Case of 12 Pack for $4,631.76
13-660Q
kimble™
K370106
14.76 in. 37.5cm 6mL Pack of 6 Each for $355.00
Case of 12 Pack for $4,260.00
13-660T
kimble™
K370108
15.74 in. 40cm 8mL Pack of 6 Each for $365.65
Case of 12 Pack for $4,387.80
13-660R
kimble™
K370107
15.74 in. 40cm 7mL Pack of 6 Each for $360.83
Case of 12 Pack for $4,329.96
13-660U
kimble™
K370109
15.74 in. 40cm 9mL Pack of 6 Each for $182.78
Case of 2 Pack for $365.56
13-660W
kimble™
K3701015
17.91 in. 45.5cm 15mL Pack of 6 Each for $398.85
Case of 12 Pack for $4,786.20
13-660M
kimble™
K37010510
12.2 in. 31cm 0.5mL Pack of 6 Each for $210.05
Case of 12 Pack for $2,520.60
13-660N
kimble™
K370101510
13.18 in. 33.5cm 1.5mL Pack of 6 Each for $294.79
Case of 2 Pack for $589.58
13-660P
kimble™
37010-2510
13.77 in. 35cm 2.5mL Pack of 6 Each for $1,127.58
Case of 12 Pack for $13,530.96
13-660C
kimble™
K370103
13.77 in. 35cm 3mL Pack of 6 Each for $365.09
Case of 2 Pack for $730.18
13-660A
kimble™
37010 1
12.2 in. 31cm 1mL Pack of 6 Each for $246.94
Case of 2 Pack for $493.88
13-660B
kimble™
37010 2
13.18 in. 33.5cm 2mL Pack of 6 Each for $316.74
Case of 2 Pack for $633.48
13-660D
kimble™
K370104
13.77 in. 35cm 4mL Pack of 6 Each for $401.94
Case of 2 Pack for $803.88
13-660F
kimble™
37010 10
15.74 in. 40cm 10mL Pack of 6 Each for $431.35
Case of 2 Pack for $862.70
13-660K
kimble™
K3701050
21.45 in. 54.5cm 50mL Case of 8 Each for $867.76
13-660E
kimble™
K370105
14.37 in. 36.5cm 5mL Pack of 6 Each for $408.06
Case of 2 Pack for $816.12
13-660J
kimble™
K3701025
19.88 in. 50.5cm 25mL Case of 12 Each for $1,094.94
13-660L
kimble™
K37010100
21.65 in. 55cm 100mL Case of 6 Each for $787.56
13-660H
kimble™
K3701020
19.88 in. 50.5cm 20mL Case of 12 Each for $1,094.94
Description & Specifications
Specifications
Certifications/Compliance Certificate of graduation accuracy, ASTM E969
Class A
Material Borosilicate Glass
Product Type Serialized Volumetric Pipet
Disposable No
Tolerance ±0.01mL
  • Meet standards of former NBS circular 602 for accuracy, appearance, quality of glass, calibration lines, and inscription Both serialized and standard volumetric pipets have tempered ends calibrated “To Deliver”
  • Each serialized/certified pipet is permanently marked with an individual serial number and comes with a Certificate of Graduation Accuracy
  • The 20, 25, and 100mL pipets come with six certificates per case; 50mL pipets, eight per case
  • All other sizes are supplied with two certificates per case
Marked with “A” to certify that they are designed from ASTM™ Specification E 969, Class A requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.