Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper


Narrow-mouth solution bottle

Manufacturer: kimble™  K15097250

 View more versions of this product

Catalog No. 02-920-1B

  • / Case of 6 Each
 

Description & Specifications

Specifications

Capacity 8.45 oz., 250mL
Capacity (Metric) 250mL
Capacity (English) 8.45 oz.
Certifications/Compliance ASTM E438-92, Type I, Class A
Closure Material PTFE
Diameter (English) 2.83 in.
Includes Red PTFE flathead stopper
Height (English) 5.7 in.
Material Borosilicate Glass
Stopper Number 19
Closure Color Gray
Diameter (Metric) 72mm
Height (Metric) 145mm
Mouth Narrow
Shape Cylindrical