Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper


Narrow-mouth solution bottle

Manufacturer: kimble™  K15097100

 View more versions of this product

Catalog No. 02-920-1A

  • / Case
 

Description & Specifications

Specifications

Capacity 3.38 oz., 100mL
Capacity (Metric) 100mL
Capacity (English) 3.38 oz.
Certifications/Compliance ASTM E438-92, Type I, Class A
Closure Material PTFE
Color Red
Diameter (English) 2.08 in.
Includes Red PTFE flathead stopper
Height (English) 4.33 in.
Material Borosilicate Glass
Stopper Number 14
Closure Color Red
Diameter (Metric) 53mm
Height (Metric) 110mm
Mouth Narrow
Shape Cylindrical