missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper
Narrow-mouth solution bottle
$234.13 - $1898.71
Specifications
| Includes | Red PTFE flathead stopper |
|---|---|
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E438-92, Type I, Class A |
| Mouth | Narrow |
| Shape | Cylindrical |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Stopper Number | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Stopper Number | Price | Quantity | |||
|
02-920-1A
|
DWK Life Sciences
15097100 |
100 mL | Red | 14 |
Case for $234.13
|
|
|||
|
02-920-1B
|
DWK Life Sciences
15097250 |
250 mL | Gray | 19 |
Case of 6 Each for $1,029.32
|
|
|||
|
02-920-1C
|
DWK Life Sciences
15097500 |
500 mL | Blue | 24 |
Case of 6 Each for $1,254.55
|
|
|||
|
02-920-1D
|
DWK Life Sciences
150971000 |
1000 mL | Green | 29 |
Case of 6 Each for $1,676.74
|
|
|||
|
02-920-1E
|
DWK Life Sciences
150972000 |
2000 mL | Green | 29 |
Case of 4 Each for $1,898.71
|
|
|||
Description
Bottle necks are ground to accept flathead color-coded PTFE stoppers.
- Sloped shoulders
- Cylindrical body and flat base
Compliance
Meet ASTM™ E 438-92, Type I, Class A requirements.
Specifications
| Red PTFE flathead stopper | |
| ASTM E438-92, Type I, Class A | |
| Cylindrical |
| Borosilicate Glass | |
| Narrow | |
| PTFE |