Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper


Narrow-mouth solution bottle

Manufacturer: kimble™  K150972000

 View more versions of this product

Catalog No. 02-920-1E

  • / Case of 4 Each
 

Description & Specifications

Specifications

Closure Color Green
Height (Metric) 265mm
Shape Cylindrical
Diameter (Metric) 136mm
Mouth Narrow
Diameter (English) 5.35 in.
Stopper Number 29
Material Borosilicate Glass
Capacity (English) 67.62 oz.
Height (English) 10.43 in.
Certifications/Compliance ASTM E438-92, Type I, Class A
Includes Red PTFE flathead stopper
Capacity 67.62 oz., 2000mL
Capacity (Metric) 2000mL
Closure Material PTFE