Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper


Narrow-mouth solution bottle

Manufacturer: kimble™  K150971000

 View more versions of this product

Catalog No. 02-920-1D

  • / Case of 6 Each
 

Description & Specifications

Specifications

Closure Color Green
Diameter (Metric) 110mm
Height (Metric) 215mm
Mouth Narrow
Shape Cylindrical
Diameter (English) 4.33 in.
Closure Material PTFE
Certifications/Compliance ASTM E438-92, Type I, Class A
Stopper Number 29
Capacity (English) 33.81 oz.
Capacity (Metric) 1000mL
Material Borosilicate Glass
Capacity 33.81 oz., 1000mL
Includes Red PTFE flathead stopper
Height (English) 8.46 in.