Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles with Flathead Color-Coded PTFE standard taper Stopper


Narrow-mouth solution bottle

Manufacturer: kimble™  K15097500

 View more versions of this product

Catalog No. 02-920-1C

  • / Case of 6 Each
 

Description & Specifications

Specifications

Closure Color Blue
Mouth Narrow
Diameter (Metric) 89mm
Shape Cylindrical
Height (Metric) 180mm
Diameter (English) 3.5 in.
Closure Material PTFE
Material Borosilicate Glass
Capacity (English) 16.9 oz.
Stopper Number 24
Capacity (Metric) 500mL
Certifications/Compliance ASTM E438-92, Type I, Class A
Includes Red PTFE flathead stopper
Height (English) 7.08 in.
Capacity 16.9 oz., 500mL