Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles


For accurate rubber stopper fit

Manufacturer: kimble™  K1509320000

 View more versions of this product

Catalog No. 02-886-8H

  • / Case
 

Description & Specifications

Specifications

Mouth Narrow
Diameter (Metric) 290mm
Height (Metric) 495mm
Diameter (English) 11.41 in.
Closure Material Rubber
Material Borosilicate Glass
Diameter (English) Inner Neck 2.28 in.
Capacity (English) 676.28 oz.
Certifications/Compliance ASTM E438-92, Type I, Class A
Stopper Number 12
Height (English) 19.4 in.
Diameter (Metric) Inner Neck 58mm
Includes Tooled neck
Capacity 676.28 oz., 20000mL
Capacity (Metric) 20000mL