missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Reagent Bottles

For accurate rubber stopper fit

Manufacturer:  Kimble™ K150931000

 View more versions of this product

Catalog No. 02-886-8D


 

Specifications Description & Specifications

Specifications

33.81 oz., 1000mL
33.81 oz.
Rubber
1.06 in.
Tooled neck
Borosilicate Glass
110mm
Narrow
1000mL
ASTM E438-92, Type I, Class A
4.33 in.
27mm
7.87 in.
6
200mm