DWK Life Sciences Kimble™ KIMAX™ Media Bottles — Ungraduated
Designed for easy pouring and handling
Supplier:  DWK Life Sciences 61100250
 View more versions of this product
Catalog No. 06-421-3
Description
Description
  • Available with (Cat. No. 06-421-7) or without graduations
  • 1000mL size features finger grips molded into the side wall and is available with two GPI thread sizes
  • All bottles have screw-thread finish
  • Without closures
Specifications
Specifications
147 mm
5.7 in.
8.45 oz., 250 mL
8.45 oz.
2.7 in.
70 mm
33-430 mm
250 mL
White
Borosilicate Glass
SDS
Documents
Documents
Accessories