DWK Life Sciences Kimble™ KIMAX™ Media Bottles — Ungraduated
Designed for easy pouring and handling
Supplier: DWK Life Sciences 61100A1000
Description
- Available with (Cat. No. 06-421-7) or without graduations
- 1000mL size features finger grips molded into the side wall and is available with two GPI thread sizes
- All bottles have screw-thread finish
- Without closures
Specifications
222 mm | |
8.7 in. | |
33.8 oz., 1000 mL | |
33.8 oz. | |
4.01 in. |
102 mm | |
38-430 mm | |
1000 mL | |
White | |
Borosilicate Glass |