DWK Life Sciences Kimble™ KIMAX™ Media Bottles — Graduated
Designed for easy pouring and handling
Manufacturer: DWK Life Sciences 61110250
Description
- Available with or without (Cat. No. 06-421-1) graduations
- 1000mL size features finger grips molded into the side wall and is available with two GPI thread sizes
- All bottles have screw-thread finish
- With or without black phenolic closures with 14B rubber liners
Specifications
33-430 mm | |
70 mm | |
147 mm | |
250 mL | |
White | |
Borosilicate Glass |
No Closure | |
5.7 in. | |
8.45 oz., 250 mL | |
8.45 oz. | |
2.7 in. | |
Black |