Kimble™ KIMAX™ Long, Straight-Neck Flask with Capacity Scale
Neck accepts metal or plastic cap or cotton plug
Manufacturer:  Kimble™ K25615250
 View more versions of this product
Catalog No. 10-140-1A
Specifications Description & Specifications
Specifications
Yes
3.8cm
50 to 225mL
5.5 in.
Plain fire-polished
8.2cm
Borosilicate Glass
38mm
Metal cap, Plastic closure, Foam plug, or Cotton plug for Cell Culture applications
25mL
14.2cm
3.2 in.
250mL
Accessories