DWK Life Sciences Kimble™ KIMAX™ Long, Straight-Neck Flask with Capacity Scale
Neck accepts metal or plastic cap or cotton plug
Supplier: DWK Life Sciences 25615125
Description
- Thick-walled tubing neck and plain fire-polished finish sealed to an Erlenmeyer-shaped body
- Graduated to capacity
- Durable white ceramic enamel scale
Compliance
Designed from ASTM™ E 1404, Type IV, Class 1 requirements
Specifications
2.5 cm | |
Borosilicate Glass | |
50 to 125 mL | |
5 in. | |
Plain fire-polished | |
6.6 cm | |
25 mm |
125 mL | |
Metal cap, Plastic closure, Foam plug, or Cotton plug for Cell Culture applications | |
25 mL | |
12.5 cm | |
2.5 in. | |
Autoclavable |