DWK Life Sciences Kimble™ KIMAX™ Gooch Filtering Crucibles
Dry precipitates to constant weight at 110°C (230°F)
Manufacturer: DWK Life Sciences 28260302
Description
- At higher temperatures, heat in electric furnace
- Do not remove crucibles until temperature is reduced to about 250°C (482°F)
- Fritted disc
- Avoid exposure to sudden temperature changes
- Coarse (C), medium (M), and fine (F) porosities
Specifications
USP Type I, ASTM E438, Type I, Class A | |
3cm | |
250°C | |
5cm | |
Medium |
With Fritted discs | |
For collecting, drying and weighing precipitates and samples to be analyzed using gravimetric analysis | |
30mL | |
Borosilicate Glass |