DWK Life Sciences Kimble™ KIMAX™ Dispensing Burets
For industrial and clinical labs
Manufacturer:  DWK Life Sciences K17080F500
 View more versions of this product
Catalog No. 03-789-5B
Includes: No. 4 stopcock with PTFE plug
Description
Description
  • Reliable, well-annealed, with durable colored white scales
Specifications
Specifications
16.9 oz.
PTFE
Dispensing laboratory solvents or solutions for a variety of clinical and industrial applications
25.4 in.
Borosilicate Glass
500mL
43mm
±2.5mL
USP Type I and ASTM E438, Type I, Class A
1.6 in.
Self-lubricating PTFE stopcock, with durable colored white scales
646mm
4
16.9 oz., 500mL
5mL
Documents