Kimble™ KIMAX™ Culture Tubes
Manufacturer:  Kimble™ K45058252
 View more versions of this product
Catalog No. S30531
Description
Description
Durable Kimax glass tubes without lip, with fire-polished top rim!
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class A
No
95mL
Borosilicate Glass
25mm
White Ceramic Enamel Marking Spot
250mm
Documents