Kimble™ KIMAX™ Culture Media Flasks
Screw-thread finish
Manufacturer:  Kimble™ K265051000
 View more versions of this product
Catalog No. 10-093D
Specifications
Description
Flasks, Erlenmeyer Cell Culture; Kimax; with Cap, 38mm-430 GPI Thread Finish; 1000mL
  • Includes long caps with cemented-in white rubber liners
Description & Specifications
Specifications
Screw
12.8cm
250 to 1000mL
8.5 in.
Cap and white Rubber liner
1000mL
5 in.
Used to mix and store culture media, as well as with other culturing techniques
100mL
21.5cm
38 to 430
Borosilicate Glass