Kimble™ KIMAX™ Colorware™ Low-Form Beakers, blue
Kimax-33 glass beakers with brightly colored graduations and marking spot
Manufacturer:  Kimble™ K14000B150
 View more versions of this product
Catalog No. 02-539-15
Specifications Description & Specifications
Specifications
Borosilicate Glass
5.07 oz.
20mL
7.9cm
150mL
20 to 140mL
ASTM, E960, Type I
Matte finish marking
Dual scale
Beaker
2.24 in.
12 Case
3.11 in.
5.07 oz., 150mL
5.7cm
Yes
Bright Blue
For easy identification in the lab, reduce cross contamination
Accessories