Kimble™ KIMAX™ Colorware™ Low-Form Beakers, red
Kimax-33 glass beakers with brightly colored graduations and marking spot
Manufacturer:  Kimble™ K14000R150
 View more versions of this product
Catalog No. 10-200-10
Specifications Description & Specifications
Specifications
Borosilicate Glass
5.07 oz.
2.24 in.
20 to 140mL
7.9cm
5.07 oz., 150mL
ASTM, E960, Type I
Raging Red
Matte finish marking
Beaker
20mL
12 Case
Dual scale
150mL
Yes
For easy identification in the lab, reduce cross contamination
5.7cm
3.11 in.
Accessories