Kimble™ KIMAX™ Colorware™ Low-Form Beakers, blue
Kimax-33 glass beakers with brightly colored graduations and marking spot
$97.69 - $183.58
Specifications
Certifications/Compliance ASTM, E960, Type I
Material Borosilicate Glass
Product Type Beaker
For Use With (Application) For easy identification in the lab, reduce cross contamination
Graduated Yes
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-539-15 Accessories Kimble™
K14000B150
150mL Case of 12 Each for $130.44
10-200 Accessories Kimble™
K14000B50
50mL Case of 12 Each for $129.14
10-200-12 Accessories Kimble™
K14000B250
250mL Case of 12 Each for $130.44
10-200-16 Accessories Kimble™
K14000B400
400mL Case of 12 Each for $153.33
10-200-20 Accessories Kimble™
K14000B600
600mL Case of 6 Each for $97.69
10-200-24 Accessories Kimble™
K14000B1000
1000mL Case of 6 Each for $183.58
10-200-48 Accessories Kimble™
K14000B100
100mL Case of 12 Each for $136.31
Specifications
Description
  • With pouring spout
  • Matte-finish marking spot can be marked with ordinary pencil
  • Colors make graduated scale easy to read
Compliance
Designed from ASTM™ E 960 Type I requirements
Description & Specifications
Specifications
ASTM, E960, Type I
Beaker
Yes
Dual scale
Borosilicate Glass
For easy identification in the lab, reduce cross contamination
Matte finish marking