Kimble™ KIMAX™ Brand Colorware Volumetric Class A Flasks
Manufacturer:  Kimble™ K28014Y50
 View more versions of this product
Catalog No. 10-214
Specifications
Description
Flask, Volumetric; Kimax; Colorware; Class A; Capacity: 50mL; Color: sunny yellow; 6/Cs.
  • Calibrated to contain with Class A accuracy but without serial number
  • Standard taper ground glass stopper is provided
  • Designed from ASTM Specification E288, Class A unserialized requirements
Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
9
Yes
14.8cm
50mL
Glass
Sunny Yellow
5.8 in.
±0.05mL
Borosilicate Glass