missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Brand Chromatography Column

Leak proof fit with all PTFE stopcock

Manufacturer:  Kimble™ 1780019300

 View more versions of this product

Catalog No. 10-310-36


 

Specifications

Description

Column, Chromatography; Kimble Chase Kontes; Kimax; Leak proof fit with all PTFE stopcock; Reinforced top; Tooled lower end; O-ring added; Liquid tight seal over 0deg.C to 100deg.C; I.D.: 19mm; Length: 300mm; Capacity: 80mL; 1/case

  • Reinforced top
  • Tooled lower end
  • O-ring added
  • Liquid tight seal over 0 to 100°C

Description & Specifications

Specifications

0 to 100°C
80mL
Glass
PTFE stopcock assembly
Chromatography Column
300mm
19mm