missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Aspirator Bottles with Tubulation

Bottom glass hose-connection outlet is fused to bottle

Manufacturer:  Kimble™ K14607500

 View more versions of this product

Catalog No. 02-973-1B


 

Requires : Tubing

Specifications Description & Specifications

Specifications

0.125 gal.
0.125 gal., 500mL
0.312 in.
22mm
89mm
6.5 in.
Yes
Narrow Mouth
Bottom hose outlet
500mL
0.86 in.
3.5 in.
7.92mm
Designed to store and discharge liquids via a bottom hose outlet
165mm
Borosilicate Glass
4
Reservoir Bottle
Accessories