DWK Life Sciences Kimble™ KIMAX™-51 Reusable Serological Pipets with Standard Opening
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Manufacturer:  DWK Life Sciences K3703311100
 View more versions of this product
Catalog No. 13-674-32E
Specifications
Specifications
Yes
0.03 oz., 1mL
Yellow
Yes
350mm
±0.02mL
1mL
0.03 oz.
ASTM E1044
Yellow
0.01mL
Tempered
Bacteriological Pipet
Borosilicate Glass
Documents