DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Manufacturer:  DWK Life Sciences K37034A25
 View more versions of this product
Catalog No. 13-674-35K
Description
Description
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Specifications
Specifications
ASTM E1044
White
0.1mL
400mm
Serological Pipet
Borosilicate Glass
White
Yes
15.74 in.
±0.1mL
25mL
Documents