DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Rugged construction
Manufacturer:  DWK Life Sciences K370202
 View more versions of this product
Catalog No. 13-664E
Description
Description
Rugged construction
  • Intended for chemical laboratory work—tip openings are smaller than usually desired for clinical laboratory operations
  • Permanent brown, graduated scale fused into uniform bore tubing ending above taper
  • Color coded by size for ease in sorting
  • Tempered tips
  • Calibrated “To Deliver”
Compliance
Designed from ASTM™ E 1293, Class B, Style 1 requirements.
Specifications
Specifications
To Deliver
0.06 oz., 2mL
B
Green
Yes
13.7 in.
±0.02mL
2mL
0.06 oz.
ASTM E438, USP Type I, Class B
Green
No
0.1mL
350mm
Mohr Pipet
Borosilicate Glass
Documents