missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets

Rugged construction

Manufacturer:  DWK Life Sciences K37020110

 View more versions of this product

Catalog No. 13-664A



Description

Description

Rugged construction

  • Intended for chemical laboratory work—tip openings are smaller than usually desired for clinical laboratory operations
  • Permanent brown, graduated scale fused into uniform bore tubing ending above taper
  • Color coded by size for ease in sorting
  • Tempered tips
  • Calibrated “To Deliver”

Compliance

Designed from ASTM™ E 1293, Class B, Style 1 requirements.

Specifications

Specifications

0.003 oz.
ASTM E438, USP Type I, Class B
White
No
0.01mL
300mm
Mohr Pipet
Borosilicate Glass
0.003 oz., 0.1mL
B
White
Yes
11.8 in.
±0.005mL
0.1mL
Documents