missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ KIMAX™-35 Milk Dilution Bottles

Designed from requirements for milk dilution bottles given in APHA Standard Methods for the Examination of Dairy Products

Manufacturer:  Kimble™ K14925160

 View more versions of this product

Catalog No. 03-422-11


 

Specifications Description & Specifications

Specifications

5 oz., 160mL
5 oz.
For milk dilution
Graduated Glass
28-400
Screw
160mL
Phenolic
5.9 in.
Black
150mm