DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures
Available in natural (clear) or choice of four colors
Manufacturer:  DWK Life Sciences K7366513
 View more versions of this product
Catalog No. 14-957-92C
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Caps for 38mm O.D. are designed for baffled culture flasks.
Specifications
Specifications
Yes
Polypropylene
Yes
Blue
Closure
13mm O.D. plain-end culture tubes
Documents