Please login to your online account to display your discounted pricing

Kick-Net

Strong net designed for rapid bioassessments.

$149.55 - $149.55

Specifications

For Use With (Application) Performing USEPA rapid bioassessment protocols for benthic macroinvertebrates
Grade Student
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S85005P

 
fisher science education
0021-P
Profesional kick net with poles Each for $149.55
Description & Specifications

Specifications

For Use With (Application) Performing USEPA rapid bioassessment protocols for benthic macroinvertebrates
Grade Student

  • Use this net for performing USEPA Rapid Bioassessment Protocols for benthic macroinvertebrates
  • 1 × 1m2, 500μm, tan net
  • Hemmed sides hold poles
  • Reinforced bottom seam helps anchoring