missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ki-67 Rat anti-Mouse, Rat, eFluor 450, Clone: SolA15, eBioscience™

Rat Monoclonal Antibody

$170.28 - $443.52

Specifications

Antigen Ki-67
Clone SolA15
Host Species Rat
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124170 Life Technologies
LS48569880
25μg Each for $170.28
501124171 Life Technologies
LS48569882
100μg Each for $443.52
Specifications

Description

The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the “Ki-67 labeling index” refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin).

The SolA15 antibody also recognizes human, non-human primate and canine Ki-67.
Description & Specifications

Specifications

Ki-67
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SolA15
Flow Cytometry (Intracellular Staining)
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL