Please login to your online account to display your discounted pricing

Ki-67 Rat anti-Mouse, Rat, Clone: SolA15, eBioscience™

Rat Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen Ki-67
Clone SolA15
Host Species Rat
Species Reactivity Canine, Cynomolgus, Human, Mouse, Non-human primate, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245563

 
life technologies
LS14569880
25μg Each for $129.05

50245564

 
life technologies
LS14569882
100μg Each for $317.55
Description & Specifications

Specifications

Antigen Ki-67
Clone SolA15
Host Species Rat
Species Reactivity Canine, Cynomolgus, Human, Mouse, Non-human primate, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the “Ki-67 labeling index” refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin).

The SolA15 antibody also recognizes human, non-human primate and canine Ki-67.