missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ki-67, Alexa Fluor 700, clone: SolA15, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS56569880

 View more versions of this product

Catalog No. 501124689



Description

Description

Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with the euchromatin at the perichromosomal layer with centromeric heterochromatin, and with the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution, with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis.

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with euchromatin at the perichromosomal layer, the centromeric heterochromatin, and the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis. Ki-67 antibodies are useful in establishing the cell growing fraction in neoplasms. In neoplastic tissues, the prognostic value is comparable to the tritiated thymidine-labelling index. The correlation between low Ki-67 index and histologically low-grade tumors is strong. Ki-67 is routinely used as a neuronal marker of cell cycling and proliferation.
Specifications

Specifications

Ki-67
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P46013, E9PVX6
antigen identified by monoclonal antibody Ki-67; antigen KI-67; KIA; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Cynomolgus Monkey, Human, Mouse, Non-human primate, Rat
Flow Cytometry
SolA15
Alexa Fluor® 700
MKI67
Liquid
MKI67
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100686578, 102135895, 17345, 291234, 4288
Documents

For Research Use Only.