Please login to your online account to display your discounted pricing

Ki-67 Mouse, eFluor 506, Clone: 20Raj1, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen Ki-67
Clone 20Raj1
Host Species Mouse
Species Reactivity Canine , Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124971

 
life technologies
LS69569941
25 tests Each for $158.05

501124972

 
life technologies
LS69569942
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen Ki-67
Clone 20Raj1
Host Species Mouse
Species Reactivity Canine , Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 506
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The monoclonal antibody 20Raj1 recognizes the human Ki-67 protein. Two isoforms of Ki-67 exist, a 345 and 395kDa form that are expressed in dividing cells. Ki-67 is expressed in all cell types and is detectable during active phases of the cell cycle (G1, S, G2, and mitosis) but is absent from resting cells (G0). During interphase, Ki-67 expression is localized to the nucleus but redistributes to the chromosomes during mitosis and has specifically been found to associate with heterochromatin-bound proteins such as chromobox protein homolog 3 (CBX3). In studies of tumor cells, Ki-67 expression has been used as a marker for determining the fraction of proliferating cells within a given population of tumor cells.

This monoclonal antibody 20Raj1 recognizes canine Ki-67.