missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ki-67 Mouse anti-Human, PE-Cy7, Clone: 20Raj1, eBioscience™

Mouse Monoclonal Antibody

$154.44 - $389.40

Specifications

Antigen Ki-67
Clone 20Raj1
Host Species Murine
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123380 Life Technologies
LS25569941
25 Tests Each for $154.44
501123381 Life Technologies
LS25569942
100 Tests Each for $389.40
Specifications

Description

The monoclonal antibody 20Raj1 recognizes the human Ki-67 protein. Two isoforms of Ki-67 exist, a 345 and 395kDa form that are expressed in dividing cells. Ki-67 is expressed in all cell types and is detectable during active phases of the cell cycle (G1, S, G2, and mitosis) but is absent from resting cells (G0). During interphase, Ki-67 expression is localized to the nucleus but redistributes to the chromosomes during mitosis and has specifically been found to associate with heterochromatin-bound proteins such as chromobox protein homolog 3 (CBX3). In studies of tumor cells, Ki-67 expression has been used as a marker for determining the fraction of proliferating cells within a given population of tumor cells.

This monoclonal antibody 20Raj1 recognizes canine Ki-67.
Description & Specifications

Specifications

Ki-67
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
20Raj1
Flow Cytometry (Intracellular Staining)
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test