Please login to your online account to display your discounted pricing

Ki-67 Mouse anti-Human, Biotin, Clone: 20Raj1, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $452.40

Specifications

Antigen Ki-67
Clone 20Raj1
Host Species Mouse
Species Reactivity Canine, Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122555

 
life technologies
LS13569982
100μg Each for $452.40

501122554

 
life technologies
LS13569980
25μg Each for $175.45
Description & Specifications

Specifications

Antigen Ki-67
Clone 20Raj1
Host Species Mouse
Species Reactivity Canine, Human
Applications Flow Cytometry (Intracellular Staining)
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 20Raj1 recognizes the human Ki-67 protein. Two isoforms of Ki-67 exist, a 345 and 395kDa form that are expressed in dividing cells. Ki-67 is expressed in all cell types and is detectable during active phases of the cell cycle (G1, S, G2, and mitosis) but is absent from resting cells (G0). During interphase, Ki-67 expression is localized to the nucleus but redistributes to the chromosomes during mitosis and has specifically been found to associate with heterochromatin-bound proteins such as chromobox protein homolog 3 (CBX3). In studies of tumor cells, Ki-67 expression has been used as a marker for determining the fraction of proliferating cells within a given population of tumor cells.

This monoclonal antibody 20Raj1 recognizes canine Ki-67.