Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Ki-67 Ab-4, Rabbit Polyclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Ki-67 Ab-4, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

$237.16 - $1,009.90

Specifications

Antigen Ki-67 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1510P

 
thermo scientific™ lab vision™
RB1510P
Purified Ab with BSA and Azide 1mL Each for $939.12

RB1510P0

 
thermo scientific™ lab vision™
RB1510P0
Purified Ab with BSA and Azide 100μL Each for $237.16

RB1510P1

 
thermo scientific™ lab vision™
RB1510P1
Purified Ab with BSA and Azide 500μL Each for $662.22

RB1510P1ABX

 
thermo scientific™ lab vision™
RB1510P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $740.16

RB1510PABX

 
thermo scientific™ lab vision™
RB1510PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,009.90

RB1510R7

 
thermo scientific™ lab vision™
RB1510R7
Ready-to-use for IHC (UVLP) 7mL Each for $648.61
Description & Specifications

Specifications

Antigen Ki-67 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen A synthetic peptide from the human Ki67 protein

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Ki-67 Ab-4, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

Ki67 is a nuclear protein, which is expressed in the proliferating cells. Ki67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiscent) phase are negative for this protein.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Middle of Ki-67 protein

Immunogen: A synthetic peptide from the human Ki-67 protein

Positive Control: Tonsil or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO